missing translation for 'onlineSavingsMsg'
Learn More

Zinc Cyanide, Technical, Approx. 85%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor Z106625KG

 View more versions of this product

Catalog No. 18614743

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

557-21-1
2.5 kg
Technical
1
∼85%
Poly Bottle

Chemical Identifiers

557-21-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.