accessibility menu, dialog, popup

UserName

ZnCNCN

CAS RN 557-21-1

IUPAC Name: zinc cyanide
Synonyms: zinc cyanide
Molecular Weight (g/mol): 117.42
Molecular Formula: C2N2Zn
InChi Key: GTLDTDOJJJZVBW-UHFFFAOYSA-N
SMILES: [Zn++].[C-]#N.[C-]#N
Percent Purity 
  • (5)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (6)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)