missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Cyanide, Technical, Approx. 85%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 25 g |
| Percent Purity | ∼85% |
| Grade | Technical |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.