missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride 98.0+%, TCI America™
Supplier: TCI America Z0014300G
Specifications
| 732°C | |
| 290°C | |
| 7646-85-7 | |
| 300 g | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| Cl[Zn]Cl | |
| 136.28 | |
| CHEBI:49976 | |
| ≥98.0% (T) |
| Zinc Chloride | |
| White | |
| Crystalline Powder | |
| Cl2Zn | |
| 2331 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| dichlorozinc | |
| 5727 | |
| 136.28 |
Chemical Identifiers
| 7646-85-7 | |
| 136.28 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| 5727 | |
| dichlorozinc |
| Cl2Zn | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| CHEBI:49976 | |
| Cl[Zn]Cl |
Safety and Handling
EINECSNumber : (1)-0264
RTECSNumber : ZH1400000
TSCA : Yes