missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride 98.0+%, TCI America™
$55.92 - $82.02
Chemical Identifiers
| CAS | 7646-85-7 |
|---|---|
| Molecular Formula | Cl2Zn |
| Molecular Weight (g/mol) | 136.28 |
| MDL Number | MFCD00011295 |
| InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
| Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
| PubChem CID | 5727 |
| ChEBI | CHEBI:49976 |
| IUPAC Name | dichlorozinc |
| SMILES | Cl[Zn]Cl |
Chemical Identifiers
| 7646-85-7 | |
| 136.28 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| 5727 | |
| dichlorozinc |
| Cl2Zn | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| CHEBI:49976 | |
| Cl[Zn]Cl |
Specifications
| 732°C | |
| White | |
| Crystalline Powder | |
| Cl2Zn | |
| 2331 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| dichlorozinc | |
| 5727 | |
| 136.28 | |
| Zinc Chloride |
| 290°C | |
| 7646-85-7 | |
| 25 g | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| Cl[Zn]Cl | |
| 136.28 | |
| CHEBI:49976 | |
| ≥98.0% (T) |
Safety and Handling
EINECSNumber : (1)-0264
RTECSNumber : ZH1400000
TSCA : Yes