missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Chloride, puriss., Honeywell Fluka™
Puriss., meets analytical specification of BP, Ph. Eur., USP, 98-100.5%
$659.97 - $3484.29
Chemical Identifiers
| CAS | 7646-85-7 |
|---|---|
| Molecular Formula | Cl2Zn |
| Molecular Weight (g/mol) | 136.28 |
| MDL Number | MFCD00011295 |
| InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
| Synonym | zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride |
| PubChem CID | 5727 |
| ChEBI | CHEBI:49976 |
| IUPAC Name | dichlorozinc |
| SMILES | Cl[Zn]Cl |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7646-85-7 | |
| 136.28 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| 5727 | |
| dichlorozinc |
| Cl2Zn | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| CHEBI:49976 | |
| Cl[Zn]Cl |
Specifications
| 7646-85-7 | |
| Cl2Zn | |
| MFCD00011295 | |
| zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride | |
| Cl[Zn]Cl | |
| 136.28 | |
| CHEBI:49976 | |
| 98 to 100.5% | |
| 1mmHg (428°C) |
| 1 kg | |
| ZnCl2 | |
| 2331 | |
| JIAARYAFYJHUJI-UHFFFAOYSA-L | |
| dichlorozinc | |
| 5727 | |
| 136.3g/mol | |
| puriss. | |
| Zinc Chloride, Meets analytical specification of Ph. Eur.BP, USP |
Safety and Handling
P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H302-H314-H410
DOTInformation : 8 / PGIII
EINECSNumber : 231-592-0
RTECSNumber : ZH1400000