missing translation for 'onlineSavingsMsg'
Learn More

Zinc Chloride, puriss., Honeywell Fluka™

Catalog No. 6001780
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001780 1 kg
60044857 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001780 Supplier Honeywell Chemicals Supplier No. 144221KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, 98-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Quantity 1 kg
Linear Formula ZnCl2
UN Number 2331
Formula Weight 136.3g/mol
Percent Purity 98 to 100.5%
Grade puriss.
Vapor Pressure 1mmHg (428°C)
Chemical Name or Material Zinc Chloride, Meets analytical specification of Ph. Eur.BP, USP
Hazard Statement P260-P280-P301 + P312 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H302-H314-H410
EINECSNumber 231-592-0
RTECSNumber ZH1400000