missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Solution , For Sulfide, Certified, 2.0N ±0.1N, LabChem™

Catalog No. p-5379534
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC271007 125 mL
LC271001 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC271007 Supplier LabChem Supplier No. LC271007
Only null left

Chemical Identifiers

CAS 5970-45-6 , 7732-18-5
Molecular Formula C4H10O6Zn
Molecular Weight (g/mol) 219.498
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Synonym zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]

Specifications

Color Colorless
CAS Max % 78, 22
Linear Formula Zn(C2H3O2)2·2H2O
Concentration 2.0N ±0.1N
Density 1.1g/mL
Solubility Information Soluble in water
Formula Weight 219.49
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Carbon monoxide; Carbon dioxide
Quantity 125 mL
Physical Form Liquid
Chemical Name or Material Zinc Acetate Solution, For Sulfide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature