missing translation for 'onlineSavingsMsg'
Learn More

Zinc Acetate Solution , For Sulfide, Certified, 2.0N ±0.1N, LabChem™

Supplier:  LabChem LC271001

 View more versions of this product

Catalog No. LC271001


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Zinc Acetate Solution, 2N
For Sulfide
78,22
Zn(C2H3O2)2·2H2O
1.1g/mL
Soluble in water
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
219.498
219.49
Passes Test
1.1g/mL
Carbon monoxide; Carbon dioxide
Liquid
Colorless
5970-45-6,7732-18-5
C4H10O6Zn
2.0N ± 0.1N
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc;diacetate;dihydrate
2724192
Certified
Poly Bottle
Traceable to NIST
500 mL

Chemical Identifiers

5970-45-6
219.498
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
zinc;diacetate;dihydrate
C4H10O6Zn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
2724192
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions