missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Zenix™ SEC-300 Gel Filtration Guard Column, 3 μm Particle Size, Stainless Steel

Catalog No. 111007761
Change view
Click to view available options
Diameter (Metric) Inner:
1 mm
2.1 mm
4.6 mm
7.8 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111007761 1 mm
111007762 2.1 mm
111007763 7.8 mm
111008004 4.6 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111007761 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777021

Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.

Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.

Specifications

pH 2.0 to 8.5
Particle Size 3 μm
Length (Metric) 5 cm
Pore Size 300 Å
Type SEC HPLC Guard Column
Diameter (Metric) Inner 1 mm
Material Stainless Steel
Phase Size Exclusion
For Use With (Application) HPLC
Product Line Zenix

Zenix is a registered trademark of Sepax Technologies