missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Zenix™ SEC-150 Gel Filtration HPLC Column, 3 μm Particle Size, PEEK

Catalog No. 111005997
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111005997 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111005997 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777020

Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.

Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 30 cm
Particle Size 3 μm
pH 2.0 to 8.5
Pore Size 150 Å
Stationary Phase Hydrophilic Polymer
Product Line Zenix
Matrix Hydrophilic Polymer Phase/Spherical Silica
Phase Size Exclusion
Material PEEK
Technique SEC HPLC
Show More Show Less

Zenix is a registered trademark of Sepax Technologies