missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Zenix™ SEC-100 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel
Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.
Supplier: MilliporeSigma™ Supelco™ Z777008
Description
Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.Specifications
| 7.8 mm | |
| 3 μm | |
| 100 Å | |
| Zenix | |
| Size Exclusion | |
| SEC HPLC |
| 30 cm | |
| 2.0 to 8.5 | |
| Hydrophilic Polymer | |
| Hydrophilic Polymer Phase/Spherical Silica | |
| Stainless Steel |
Zenix is a registered trademark of Sepax Technologies