missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Zenix™-C SEC-300 Gel Filtration Guard Column, 3 μm Particle Size, Stainless Steel
The bonded phase in Zenix™-C SEC columns is more polar than the hydrophilic phase used in Zenix™ columns, making Zenix™-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies.
$1192.97 - $1192.97
Specifications
| pH | 2.0 to 8.5 |
|---|---|
| Particle Size | 3 μm |
| Length (Metric) | 5 cm |
| Pore Size | 300 Å |
| Type | SEC HPLC Guard Column |
Description
Both Zenix™-C and Zenix™ columns are based on the same high efficiency 3 micron particles and each product line is available in the same wide range of pore sizes to tailor the optimal pore size to the hydrodynamic radius of the biopolymer of interest.Specifications
| 2.0 to 8.5 | |
| 5 cm | |
| SEC HPLC Guard Column | |
| Size Exclusion | |
| Zenix |
| 3 μm | |
| 300 Å | |
| Stainless Steel | |
| HPLC |
Zenix is a registered trademark of Sepax Technologies