missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Zenix™-C SEC-100 Gel Filtration Guard Column, 3 μm Particle Size, Stainless Steel

Catalog No. 111007764
Change view
Click to view available options
Diameter (Metric) Inner:
7.8 mm
3 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111007764 3 m
111007671 7.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111007764 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777072

The bonded phase in Zenix™-C SEC columns is more polar than the hydrophilic phase used in Zenix™ columns, making Zenix™-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies.

Both Zenix™-C and Zenix™ columns are based on the same high efficiency 3 micron particles and each product line is available in the same wide range of pore sizes to tailor the optimal pore size to the hydrodynamic radius of the biopolymer of interest.

Specifications

pH 2.0 to 8.5
Particle Size 3 μm
Length (Metric) 5 cm
Pore Size 100 Å
Type SEC HPLC Guard Column
Diameter (Metric) Inner 3 m
Material Stainless Steel
Phase Size Exclusion
For Use With (Application) HPLC
Product Line Zenix

Zenix is a registered trademark of Sepax Technologies