Learn More
Zaprinast, 98+%
CAS: 37762-06-4 | C13H13N5O2 | 271.28 g/mol
Supplier: Thermo Scientific Chemicals J63326MF
Description
Phosphodiesterase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Zaprinast | |
| Glass bottle | |
| MFCD00214073 | |
| zaprinast, zaprinastum inn-latin, unii-gxt25d5ds0, 8-aza-2-2-propoxyphenyl-6-purinone, tocris-0947, 2-o-propoxyphenyl-8-azapurine-6-one, 2-o-propoxyphenyl-8-azapurin-6-one, gxt25d5ds0, 2-2-propoxyphenyl-8-aza-6-purinone, 5-2-propoxyphenyl-1h-1,2,3 triazolo 4,5-d pyrimidin-7-ol | |
| CCCOC1=CC=CC=C1C2=NC(=O)C3=NNNC3=N2 | |
| 271.28 | |
| 271.28 | |
| Powder |
| 37762-06-4 | |
| C13H13N5O2 | |
| 50 mg | |
| REZGGXNDEMKIQB-UHFFFAOYSA-N | |
| 5-(2-propoxyphenyl)-2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one | |
| 5722 | |
| ≥98% |
Chemical Identifiers
| 37762-06-4 | |
| 271.28 | |
| REZGGXNDEMKIQB-UHFFFAOYSA-N | |
| 5722 | |
| CCCOC1=CC=CC=C1C2=NC(=O)C3=NNNC3=N2 |
| C13H13N5O2 | |
| MFCD00214073 | |
| zaprinast, zaprinastum inn-latin, unii-gxt25d5ds0, 8-aza-2-2-propoxyphenyl-6-purinone, tocris-0947, 2-o-propoxyphenyl-8-azapurine-6-one, 2-o-propoxyphenyl-8-azapurin-6-one, gxt25d5ds0, 2-2-propoxyphenyl-8-aza-6-purinone, 5-2-propoxyphenyl-1h-1,2,3 triazolo 4,5-d pyrimidin-7-ol | |
| 5-(2-propoxyphenyl)-2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 253-655-1
RTECSNumber : XZ6157358
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only