accessibility menu, dialog, popup

UserName

CAS RN 37762-06-4

IUPAC Name: 5-(2-propoxyphenyl)-2H,3H,7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one
Synonyms: zaprinast
Molecular Weight (g/mol): 271.28
Molecular Formula: C13H13N5O2
InChi Key: REZGGXNDEMKIQB-UHFFFAOYSA-N
SMILES: CCCOC1=CC=CC=C1C1=NC(=O)C2=NNNC2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results