missing translation for 'onlineSavingsMsg'
Learn More

Zaprinast, 98+%

Catalog No. p-7076609
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
AAJ63326MA 10 mg
AAJ63326MF 50 mg
2 options

Catalog No. AAJ63326MA

Supplier: Thermo Scientific Chemicals J63326MA

Only null left

CAS: 37762-06-4 | C13H13N5O2 | 271.28 g/mol

Phosphodiesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37762-06-4
Molecular Formula C13H13N5O2
Molecular Weight (g/mol) 271.28
MDL Number MFCD00214073
InChI Key REZGGXNDEMKIQB-UHFFFAOYSA-N
Synonym zaprinast, zaprinastum inn-latin, unii-gxt25d5ds0, 8-aza-2-2-propoxyphenyl-6-purinone, tocris-0947, 2-o-propoxyphenyl-8-azapurine-6-one, 2-o-propoxyphenyl-8-azapurin-6-one, gxt25d5ds0, 2-2-propoxyphenyl-8-aza-6-purinone, 5-2-propoxyphenyl-1h-1,2,3 triazolo 4,5-d pyrimidin-7-ol
PubChem CID 5722
IUPAC Name 5-(2-propoxyphenyl)-2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
SMILES CCCOC1=CC=CC=C1C2=NC(=O)C3=NNNC3=N2

Specifications

Packaging Glass bottle
Quantity 10 mg
Formula Weight 271.28
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Zaprinast
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-655-1
RTECSNumber XZ6157358
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only