missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Zanamivir
Description
Zanamivir is an influenza viral neuraminidase inhibitor (IC50 values are 0.95 nM and 2.7 nM for influenza A and B, respectively). Inhibits neuraminidase, preventing the cleavage of sialic acid on the cell receptors, and release of the newly formed virions.
Specifications
Specifications
| Chemical Name or Material | 5-(Acetylamino)-4-[(aminoiminomethyl)amino]-2,6-anhydro-3,4,5-trideoxy-5-(Acetylamino)-4-[(aminoiminomethyl)amino]-2,6-anhydro-3,4,5-trideoxy-D-glycero-D-galactonon-2-enonic acid-glycero-D-galactonon-2-enonic acid |
| CAS | 139110-80-8 |
| Quantity | 50 mg |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C12H20N4O7 |
| Purity | 0.98 |