Learn More
Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis)
CAS: 252976-51-5 | C3F9O9S3Yb | 620.23 g/mol
Supplier: Thermo Scientific Chemicals 304170050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
| Ytterbium(III) triflate hydrate | |
| White | |
| Crystalline Powder | |
| 99.90% | |
| (CF3SO3)3Yb·xH2O | |
| ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate | |
| AHZJKOKFZJYCLG-UHFFFAOYSA-K | |
| ytterbium(3+) tritrifluoromethanesulfonate | |
| 22836430 | |
| 99.9% | |
| Glass bottle |
| 118.0°C to 122.0°C | |
| 252976-51-5 | |
| 5 g | |
| C3F9O9S3Yb | |
| MFCD06200261 | |
| Solubility in water: soluble | |
| [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F | |
| 620.23 | |
| 620.23 | |
| Trace Metal Basis |
Chemical Identifiers
| 252976-51-5 | |
| 620.23 | |
| AHZJKOKFZJYCLG-UHFFFAOYSA-K | |
| 22836430 | |
| [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
| C3F9O9S3Yb | |
| MFCD06200261 | |
| ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate | |
| ytterbium(3+) tritrifluoromethanesulfonate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
RUO – Research Use Only