missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis)

Catalog No. p-3777103
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC304170010 1 g Glass bottle
AC304170050 5 g Glass bottle
AC304170250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC304170010 Supplier Thermo Scientific Chemicals Supplier No. 304170010
Only null left

CAS: 252976-51-5 | C3F9O9S3Yb | 620.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 252976-51-5
Molecular Formula C3F9O9S3Yb
Molecular Weight (g/mol) 620.23
MDL Number MFCD06200261
InChI Key AHZJKOKFZJYCLG-UHFFFAOYSA-K
Synonym ytterbium iii trifluoromethanesulfonate hydrate, ytterbium 3+ hydrate tritriflate, ytterbium iii triflate hydrate, ytterbium triflate hydrate, acmc-1aqes, yb otf 3.h2o, ytterbium triflate monohydrate, ytterbium trifluoromethanesulfonate hydrate, ytterbium trifluoromethanesulfonate, hydrate
PubChem CID 22836430
IUPAC Name ytterbium(3+) tritrifluoromethanesulfonate
SMILES [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 118.0°C to 122.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 99.90%
Linear Formula (CF3SO3)3Yb·xH2O
Solubility Information Solubility in water: soluble
Formula Weight 620.23
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) triflate hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doc

RUO – Research Use Only