CAS RN 252976-51-5
IUPAC Name:
ytterbium(3+) tritrifluoromethanesulfonate
Synonyms:
ytterbium triflate
Molecular Weight (g/mol):
620.23
Molecular Formula:
C3F9O9S3Yb
InChi Key:
AHZJKOKFZJYCLG-UHFFFAOYSA-K
SMILES:
[Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
Ytterbium(III) triflate hydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals
CAS: 252976-51-5 Molecular Formula: C3F9O9S3Yb Molecular Weight (g/mol): 620.23 MDL Number: MFCD06200261 InChI Key: AHZJKOKFZJYCLG-UHFFFAOYSA-K Synonym: ytterbium iii trifluoromethanesulfonate hydrate,ytterbium 3+ hydrate tritriflate,ytterbium iii triflate hydrate,ytterbium triflate hydrate,acmc-1aqes,yb otf 3.h2o,ytterbium triflate monohydrate,ytterbium trifluoromethanesulfonate hydrate,ytterbium trifluoromethanesulfonate, hydrate PubChem CID: 22836430 IUPAC Name: ytterbium(3+) tritrifluoromethanesulfonate SMILES: [Yb+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F