missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ YNT 185
Description
YNT 185 is a potent and selective orexin OX2 receptor agonist (EC50 = 28 nM at human OX2 expressed in CHO cells). Displays approximately 100-fold selectivity for OX2 over OX1 (EC50 = 2.75 μM at human OX1 expressed in CHO cells). Depolarizes OX2-expressing histaminergic neurons in mouse brain slices. Increases wake time in wild type mice. Suppresses cataplexy-like symptoms in OX knockout mice.
Specifications
Specifications
| Chemical Name or Material | 3-[[[3-[[2-[[2-(Dimethylamino)benzoyl]amino]ethyl]amino]phenyl]amino]sulfonyl]-4-methoxy-N,N-dimethyl-[1,1-biphenyl]-3-carboxamide dihydrochloride |
| CAS | 1804978-82-2 |
| Quantity | 10 mg |
| Target | OX2 Receptor Agonists |
| Molecular Formula | C33H37N5O5S0.2 HCl |
| Formula Weight | 2778 |
| Purity | 98% |