missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ YK 4-279

Catalog No. 406710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
406710 10 mg
406750 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 406710 Supplier Tocris Bioscience™ Supplier No. 4067/10*

Inhibitor of RNA helicase A (RHA)

Inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1. Inhibits Ewing's sarcoma family tumor (ESFT) cell growth; induces apoptosis.

Chemical Identifiers

CAS 1037184-44-3
Molecular Formula C17H13Cl2NO4

Specifications

Quantity 10 mg
Formula Weight 366.2
Purity >99%
Molecular Weight (g/mol) 366.19
Chemical Name or Material YK 4-279
Recommended Storage Store at +4°C