missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Yellow Zirconium Oxide Beads
Description
- Useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing
- For isolation of RNA and Proteins from bacterial aggregates, biofilms, and whole insects
- Sample Type: Plants, Animal Tissues, Wood, Seeds, Soil, Clay, Bacteria
Specifications
Specifications
| Bead Type | Zirconium Oxide |
| Color | Yellow |
| Content And Storage | Room temperature |
| Sample Type | Plants, Animal Tissues, Wood, Seeds, Soil, Clay, Bacteria |
| For Use With (Equipment) | FastPrep-24 5G, FastPrep-24, FastPrep-96 |
| For Use With (Application) | Extractions of nucleic acids, proteins, metabolites and other small molecules |
| Product Type | Yellow Zirconium Oxide Bead |
| Quantity | 1 kg |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.