missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast Lytic Enzyme, Arthrobacter luteus

Catalog No. aaj6319503
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
250 mg
500 mg
Catalog No. Quantity
AAJ6319503 1 g
AAJ63195MD 250 mg
AAJ63195ME 500 mg
3 options

Catalog No. AAJ6319503

Supplier: Thermo Scientific Chemicals J6319503

Only null left

Hydrolyzes linear glucose polymers with beta-1,3-linkages | CAS: 37340-57-1

Yeast Lytic Enzyme is used to hydrolyzes linear glucose polymers with beta-1,3-linkages, used for enzymatic digestion of yeast glycan coats and for spheroplast formation, used to digest the cell wall of active yeast cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 37340-57-1
Synonym Lyticase; Zymolyase 20T

Specifications

Color Off-white
Product Type Cell Lysis Enzyme
Content And Storage 2°C to 8°C
Form Lyophilized
For Use With (Application) Hydrolyzes linear glucose polymers with beta-1,3-linkages
Quantity 1 g
Recommended Storage Keep cold

RUO – Research Use Only