missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yeast Extract, Powder, MP Biomedicals™
Yeast extract From Yeast, Molecular Biology Reagent, Yellow-tan Powder
$582.00 - $582.00
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19402791
|
MP Biomedicals Inc
0219402791 |
1 kg |
Each for $582.00
|
|
|||||
Description
- It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.
- Yeast extract powder is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast.
- It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
Chemical Identifiers
| 8013-01-2 |
Specifications
| 8013-01-2 | |
| Solid | |
| 1 kg |
| Tan-Yellow | |
| Molecular Biology Grade | |
| Yeast Extract |
Safety and Handling
EINECSNumber : 232-387-9