missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yeast extract

CAS: 84604-16-

$281.39 - $1607.61

Chemical Identifiers

CAS 84604-16-
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC451120010
SDS View Documents
Thermo Scientific Chemicals
451120010
1 kg
Each for $281.39
Only null left
 
AC451120050
SDS View Documents
Thermo Scientific Chemicals
451120050
5kg
Each for $796.23
Only null left
 
AC451120250
SDS View Documents
Thermo Scientific Chemicals
451120250
25 kg
Each for $1,607.61
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84604-16-

Specifications

84604-16-
Plastic Bottle
1 kg
Yeast Extract
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 283-294-5

SDS
Documents

Documents

RUO – Research Use Only