missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Y-27632 in solution
Description
Y-27632 in solution is a convenient and ready-to-use 10 mM sterile-filtered solution of Y-27632 (Cat. No. 1254), pre-dissolved in water. Increases survival rate of human embryonic stem (hES) cells and iPSC undergoing cryopreservation. Optimizes naive human pluripotent stem cell growth and viability following naive cell derivation from primed ESCs and iPSCs using naive human stem cell medium (NHSM). Used as a component of growth media for urothelial organoids and in a protocol to generate brain organoids from human iPSCs.
Supplied in vials containing a minimum of 1 mL of Y-27632 solution.
Specifications
Specifications
| CAS | 129830-38-2 |
| Quantity | 4 mg |
| Product Line | Tocris |
| Target | Rho-kinase Inhibitors |
| Molecular Formula | C14H21N3O.2HCl |
| Purity | 0.97 |