missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylometazoline Hydrochloride 98.0+%, TCI America™
$82.02 - $208.78
Chemical Identifiers
| CAS | 1218-35-5 |
|---|---|
| Molecular Formula | C16H25ClN2 |
| Molecular Weight (g/mol) | 280.84 |
| MDL Number | MFCD00238707 |
| InChI Key | YGWFCQYETHJKNX-UHFFFAOYSA-N |
| Synonym | xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride |
| PubChem CID | 5282386 |
| IUPAC Name | 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride |
| SMILES | CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl |
Chemical Identifiers
| 1218-35-5 | |
| 280.84 | |
| YGWFCQYETHJKNX-UHFFFAOYSA-N | |
| 5282386 | |
| CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl |
| C16H25ClN2 | |
| MFCD00238707 | |
| xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride | |
| 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride |
Specifications
| 1218-35-5 | |
| C16H25ClN2 | |
| 1 g | |
| xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride | |
| CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl | |
| 280.84 | |
| 280.84 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00238707 | |
| 2811 | |
| YGWFCQYETHJKNX-UHFFFAOYSA-N | |
| 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride | |
| 5282386 | |
| ≥98.0% (HPLC,N) | |
| Xylometazoline Hydrochloride |
Safety and Handling
RTECSNumber : NJ2390000
TSCA : Yes