accessibility menu, dialog, popup

UserName

HClCH3CH3NHNH3CCH3H3C

CAS RN 1218-35-5

IUPAC Name: hydrogen 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole chloride
Synonyms: xylometazoline hydrochloride
Molecular Weight (g/mol): 280.84
Molecular Formula: C16H25ClN2
InChi Key: YGWFCQYETHJKNX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC(=CC(C)=C1CC1=NCCN1)C(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)