missing translation for 'onlineSavingsMsg'
Learn More

Xylometazoline Hydrochloride 98.0+%, TCI America™

Catalog No. X00635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X00635G 5 g
X00631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X00635G Supplier TCI America Supplier No. X00635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1218-35-5
Molecular Formula C16H25ClN2
Molecular Weight (g/mol) 280.84
MDL Number MFCD00238707
InChI Key YGWFCQYETHJKNX-UHFFFAOYSA-N
Synonym xylometazoline hydrochloride, xylometazoline hcl, 2-4-tert-butyl-2,6-dimethylbenzyl-2-imidazoline hydrochloride, unii-x5s84033nz, dsstox_cid_25223, dsstox_rid_80760, dsstox_gsid_45223, 2-4-tert-butyl-2,6-dimethyl-phenyl methyl-4,5-dihydro-1h-imidazole hydrochloride, 2-4-tert-butyl-2,6-dimethylphenyl methyl-4,5-dihydro-1h-imidazole hydrochloride
PubChem CID 5282386
IUPAC Name 2-[(4-tert-butyl-2,6-dimethylphenyl)methyl]-4,5-dihydro-1H-imidazole;hydrochloride
SMILES CC1=CC(=CC(=C1CC2=NCCN2)C)C(C)(C)C.Cl

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 280.84
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Xylometazoline Hydrochloride
RTECSNumber NJ2390000
TSCA Yes