missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylitol, NF, 98.5-101%, Spectrum™ Chemical
X1017, 87-99-0, C5H12O5
$286.58 - $15726.88
Chemical Identifiers
| CAS | 87-99-0 |
|---|---|
| Molecular Formula | C5H12O5 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00064291,MFCD00064292 |
| InChI Key | HEBKCHPVOIAQTA-UHFFFAOYNA-N |
| IUPAC Name | pentane-1,2,3,4,5-pentol |
| SMILES | OCC(O)C(O)C(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18607779
|
Spectrum Chemical Mfg Cor
X1017100GM |
100 g | Poly Bottle |
Each for $286.58
|
|
||||
|
18607780
|
Spectrum Chemical Mfg Cor
X1017500GM |
500 g | Poly Bottle |
Each for $812.45
|
|
||||
|
18607781
|
Spectrum Chemical Mfg Cor
X101725KG |
2.5 kg | Poly Pail |
Each for $2,221.82
|
|
||||
|
18607782
|
Spectrum Chemical Mfg Cor
X101712KG |
12 kg | Poly Pail |
Each for $4,648.36
|
|
||||
|
18607783
|
Spectrum Chemical Mfg Cor
X101750KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Xylitol, NF is used as a sugar substitute in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 87-99-0 | |
| 152.15 | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| OCC(O)C(O)C(O)CO |
| C5H12O5 | |
| MFCD00064291,MFCD00064292 | |
| pentane-1,2,3,4,5-pentol |
Specifications
| 87-99-0 | |
| 0.005 | |
| C5H12O5 | |
| 100 g | |
| OCC(O)C(O)C(O)CO | |
| 152.15 | |
| NF |
| 1 | |
| Poly Bottle | |
| MFCD00064291,MFCD00064292 | |
| HEBKCHPVOIAQTA-UHFFFAOYNA-N | |
| pentane-1,2,3,4,5-pentol | |
| 98.5 to 101% |