missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol Orange, sodium salt, pure, water soluble

CAS: 3618-43-7 | C31H28N2Na4O13S

$99.34 - $1906.05

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211220010
SDS View Documents
Thermo Scientific Chemicals
211220010
1 g Glass bottle
Each for $99.34
Only null left
 
AC211220050
SDS View Documents
Thermo Scientific Chemicals
211220050
5 g Glass bottle
Each for $298.43
Only null left
 
AC211220100
SDS View Documents
Thermo Scientific Chemicals
211220100
10 g Glass bottle
Each for $367.07
Case of 6 Each for $0.00
Only null left
 
AC211220500
SDS View Documents
Thermo Scientific Chemicals
211220500
50 g Glass bottle
Each for $1,906.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

3618-43-7
C31H28N2Na4O13S

Specifications

210.0°C
C31H28N2Na4O13S
Authentic
10.0% (110°C, 1 hr) max.
Pure
(for Zinc titration) Passes test
Brown to Brown/Red
Crystalline Powder
3618-43-7
Solubility in water: 510g/L (20°C).
575 to 585nm (0.1N NaOH)
760.59
Glass bottle
Complies
1 g
Xylenol Orange, sodium salt
Videos
SDS
Documents

Documents

RUO – Research Use Only