missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylenol Orange, sodium salt, pure, water soluble

CAS: 3618-43-7 | C31H28N2Na4O13S

Supplier:  Thermo Scientific Chemicals 211220010

 View more versions of this product

Catalog No. AC211220010


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

Xylenol Orange, sodium salt
3618-43-7
Solubility in water: 510g/L (20°C).
575 to 585nm (0.1N NaOH)
760.59
Glass bottle
Complies
1 g
210.0°C
C31H28N2Na4O13S
Authentic
10.0% (110°C, 1 hr) max.
Pure
(for Zinc titration) Passes test
Brown to Brown/Red
Crystalline Powder

Chemical Identifiers

3618-43-7
C31H28N2Na4O13S
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only