missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Indicator, 0.2% (w/v) in Alcohol, Ricca Chemical

Supplier:  Ricca Chemical Company 938816

Quantity 500 mL
Packaging Natural Poly Bottle

 View more versions of this product

Catalog No. 938816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

3618-43-7

Specifications

Xylenol Orange
0.2% (w/v) Alcoholic Solution, in SDA-3A (100 parts Ethanol and 5 parts Methanol)
500 mL
3618-43-7
Natural Poly Bottle
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 30 days

SDS
Documents

Documents