missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Technical, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor X10034LT

 View more versions of this product

Catalog No. 18614683


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Xylenes
75 to 87%,13 to 25%
4 L
Technical
1330-20-7,100-41-4
136°C to 141°C
Amber Glass Bottle

Chemical Identifiers

1330-20-7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.