missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Technical, Spectrum™ Chemical

Catalog No. p-7166733
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614684 500 mL Amber Glass Bottle
18614683 4 L Amber Glass Bottle
18614681 20 L Steel Can
18614682 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614684 Supplier Spectrum Chemical Mfg Cor Supplier No. X1003500MLGL
Only null left

Chemical Identifiers

CAS 1330-20-7 , 100-41-4

Specifications

CAS Max % 75 to 87%, 13 to 25%
Quantity 500 mL
Packaging Amber Glass Bottle
Boiling Point 136°C to 141°C
Grade Technical
Chemical Name or Material Xylenes

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.