missing translation for 'onlineSavingsMsg'
Learn More

m-Tolyl acetate, 97%

Catalog No. AAJ61726MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61726MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61726MD Supplier Thermo Scientific Chemicals Supplier No. J61726MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 111072-31-2 | C22H16N7NaO13S2 | 673.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Tetrazolium derivative useful in cancer research.XTT sodium salt is used in conjunction with phenazine methosulfate to screen human cancer cell lines. It is also used to study fungal cell damage. It is involved in testing antimicrob

Chemical Identifiers

CAS 111072-31-2
Molecular Formula C22H16N7NaO13S2
Molecular Weight (g/mol) 673.52
MDL Number MFCD00083517
InChI Key JACYMBNQPPWQML-UHFFFAOYSA-M
Synonym xtt
PubChem CID 131668615
IUPAC Name sodium;2-(2-methoxy-4-nitro-5-sulfonatophenyl)-3-(2-methoxy-4-nitro-5-sulfophenyl)-N-phenyltetrazol-3-ium-5-carboximidate
SMILES [Na+].COC1=C(C=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O)N1N=C(N=[N+]1C1=C(OC)C=C(C(=C1)S([O-])(=O)=O)[N+]([O-])=O)C(=O)NC1=CC=CC=C1

Specifications

Color Yellow
Quantity 250 mg
Solubility Information Soluble in hot water at 2;5mg/ml;Soluble in water,dimethyl sulfoxide,dimethyl formamide and phosphate buffered saline;at pH =7;2.
Formula Weight 673.52
Physical Form Powder
Chemical Name or Material XTT sodium salt
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only