accessibility menu, dialog, popup

UserName

CAS RN 111072-31-2

IUPAC Name: sodium 2,3-bis(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium
Synonyms: sodium 2,3-bis(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)-1,2λ⁵,3,4-tetrazol-2-ylium
Molecular Weight (g/mol): 673.52
Molecular Formula: C22H16N7NaO13S2
InChi Key: JACYMBNQPPWQML-UHFFFAOYSA-M
SMILES: [Na+].COC1=C(C=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O)N1N=C(N=[N+]1C1=C(OC)C=C(C(=C1)S([O-])(=O)=O)[N+]([O-])=O)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results