missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ xMAP™ Sheath Concentrate PLUS, RUO
Description
xMAP Sheath Concentrate PLUS, RUO, is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, and xMAP INTELLIFLEX DRSE systems.
- 175 mL replacement bottle for wash solution
Specifications
Specifications
| Concentration | 20X |
| For Use With (Equipment) | Luminex 200 Instrument System, Luminex FLEXMAP 3 D Instrument System, Luminex xMAP INTELLIFLEX, Luminex xMAP INTELLIFLEX DR-SE |
| Quantity | 1 L |
For Research Use Only. Not for use in diagnostic procedures.