Learn More
Xanthene-9-carboxylic acid, 98%
CAS: 82-07-5 | C14H10O3 | 226.231 g/mol
Supplier: Thermo Scientific Chemicals L1038414
Description
Transesterification of the ethyl ester of commercially available xanthene-9-carboxylic acid yeildits 3- quinuclidinyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTransesterification of the ethyl ester of commercially available xanthene-9-carboxylic acid yeildits 3- quinuclidinyl ester.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| Xanthene-9-carboxylic acid | |
| ∽217°C (decomposition) | |
| MFCD00005059 | |
| 174414 | |
| Insoluble in water. | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)C(=O)O | |
| 226.231 | |
| 226.23 |
| 82-07-5 | |
| C14H10O3 | |
| 25 g | |
| xanthene-9-carboxylic acid, xanthanoic acid, xanthenecarboxylic acid, 9-xanthenecarboxylic acid, unii-63a81f6a8v, xanthene-9-carboxylicacid, pubchem17697, maybridge1_006340, 9-xanthenylcarboxylic acid, chembl520 | |
| VSBFNCXKYIEYIS-UHFFFAOYSA-N | |
| 9H-xanthene-9-carboxylic acid | |
| 65736 | |
| 98% |
Chemical Identifiers
| 82-07-5 | |
| 226.231 | |
| VSBFNCXKYIEYIS-UHFFFAOYSA-N | |
| 65736 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)C(=O)O |
| C14H10O3 | |
| MFCD00005059 | |
| xanthene-9-carboxylic acid, xanthanoic acid, xanthenecarboxylic acid, 9-xanthenecarboxylic acid, unii-63a81f6a8v, xanthene-9-carboxylicacid, pubchem17697, maybridge1_006340, 9-xanthenylcarboxylic acid, chembl520 | |
| 9H-xanthene-9-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 201-394-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only