accessibility menu, dialog, popup

UserName

CAS RN 82-07-5

IUPAC Name: 9H-xanthene-9-carboxylic acid
Synonyms: xanthene-9-carboxylic acid
Molecular Weight (g/mol): 226.23
Molecular Formula: C14H10O3
InChi Key: VSBFNCXKYIEYIS-UHFFFAOYSA-N
SMILES: OC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)