missing translation for 'onlineSavingsMsg'
Learn More

Xanthene-9-carboxylic acid, 98%

Catalog No. p-7045350
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1038406 5 g
AAL1038414 25 g
AAL1038422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1038406 Supplier Thermo Scientific Chemicals Supplier No. L1038406
Only null left

CAS: 82-07-5 | C14H10O3 | 226.231 g/mol

Transesterification of the ethyl ester of commercially available xanthene-9-carboxylic acid yeildits 3- quinuclidinyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Transesterification of the ethyl ester of commercially available xanthene-9-carboxylic acid yeildits 3- quinuclidinyl ester.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 82-07-5
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.231
MDL Number MFCD00005059
InChI Key VSBFNCXKYIEYIS-UHFFFAOYSA-N
Synonym xanthene-9-carboxylic acid, xanthanoic acid, xanthenecarboxylic acid, 9-xanthenecarboxylic acid, unii-63a81f6a8v, xanthene-9-carboxylicacid, pubchem17697, maybridge1_006340, 9-xanthenylcarboxylic acid, chembl520
PubChem CID 65736
IUPAC Name 9H-xanthene-9-carboxylic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C3O2)C(=O)O

Specifications

Melting Point ∽217°C (decomposition)
Quantity 5 g
Beilstein 174414
Solubility Information Insoluble in water.
Formula Weight 226.23
Percent Purity 98%
Chemical Name or Material Xanthene-9-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 201-394-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only