missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ X-Gal

Catalog No. FERR0402
Change view
Click to view available options
Quantity:
1 g
2 x 1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0402 2 x 1 g
FERR0404 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERR0402 Supplier Thermo Scientific™ Supplier No. R0402

Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.

X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.

Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.

Specifications

Detection Method Colorimetric
Product Type X-Gal
Quantity 2 x 1 g
Substrate Properties Chemical Substrate
Substrate Type Beta-Gal Substrate, X-Gal

For Research Use Only. Not for use in diagnostic procedures.