missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WYE 687 dihydrochloride

Catalog No. 428210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
428210 10 mg
428250 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 428210 Supplier Tocris Bioscience™ Supplier No. 4282/10

Potent and selective mTOR inhibitor

Potent, ATP-competitive inhibitor of mammalian target of rapamycin (mTOR) (IC50 = 7 nM). Displays selectivity for mTOR over PI 3-Kα (∽100-fold) and PI 3-Kγ (∽500-fold). Inhibits phosphorylation of mTORC1 and mTORC2 substrates including S6K, SGK and Akt; blocks VEGF secretion and HIF-1α expression. Exhibits antiproliferative effects in cancer cell lines through G1 cell cycle arrest and selective apoptosis.

Chemical Identifiers

CAS 1062161-90-3
Molecular Formula C28H32N8O3

Specifications

Quantity 10 mg
Formula Weight 601.53
Purity >98%
Molecular Weight (g/mol) 528.617
Chemical Name or Material WYE 687 dihydrochloride
Recommended Storage Desiccate at Room Temperature