missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WS 6
Description
WS 6 induces growth and proliferation of cultured islet α and β cells in vitro (EC50 = 0.28 μM at β cells). Inhibits growth of FLT3-ITD+ AmL and K562 CmL cells (IC50 values are <0.24 nM and 40.48 nM, respectively). Reduces blood glucose in an animal model of type 1 diabetes. In combination with TTNPB (Cat. No. 0761) and 1-azakenpaullone, WS 6 induces and maintains murine totipotent stem cells. In an animal model of depression WS 6 induces dendrite complexity, enhances hippocampal neurogenesis, increases GABAergic neurons and GABA expression, and displays antidepressant and anxiolytic activity. WS 6 is predicted to be a receptor tyrosine kinase inhibitor.
Specifications
Specifications
| Chemical Name or Material | 4-[[6-[(Cyclopropylcarbonyl)amino]-4-pyrimidinyl]oxy]-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]benzeneacetamide |
| CAS | 1421227-53-3 |
| Quantity | 10 mg |
| Target | Compounds for Stem Cell Proliferation and Renewal |
| Molecular Formula | C29H31F3N6O3 |
| Purity | 98% |