Learn More
Brady™ WorkHorse™ B-423 Permanent Polyester Labels
For use in commercial/industrial applications. Brady B-423 polyester label are for use with BMP71 printer.
$118.71
Specifications
Certifications/Compliance UL recognized, CSA accepted, AGA approved, Halogen free (DIN VDE 0472/part 815)
Description Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
Adhesive Type Acrylic, Permanent
Color White
Finish Gloss
View More Specs
Products 1
Catalog Number Mfr. No. Size (English) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Size (English) Quantity Price Quantity & Availability  
19109150
View Documents Promotion Details
Brady™
M715423
0.5 x 0.2 in. (W x H) 750/Roll
Each for $118.71
Only null left
Description
Description
Specifications
Specifications
UL recognized, CSA accepted, AGA approved, Halogen free (DIN VDE 0472/part 815)
Acrylic, Permanent
Gloss
Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates
0.003 in.
Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
White
Polyester (B-423)
BMP™71 Printer