missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT5A Recombinant Rabbit Monoclonal Antibody (23GB2400)

Rabbit Recombinant Monoclonal Antibody
Specifications
| Antigen | WNT5A |
|---|---|
| Clone | 23GB2400 |
| Concentration | 0.4 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
Description
WNT5A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.Specifications
| WNT5A | |
| 0.4 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P41221 | |
| 7474 | |
| A synthesized peptide derived from human Wnt5a (330-360AA). | |
| Antibody |
| 23GB2400 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| WNT5A | |
| 8030457G12Rik; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; Wnt family member 5A; WNT5A; Wnt-5a; WNT-5A protein | |
| WNT5A | |
| Primary | |
| Affinity chromatography |