missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT5A Recombinant Rabbit Monoclonal Antibody (23GB2400)

Catalog No. MA553583
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553583 20 μL
MA553582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553583 Supplier Invitrogen™ Supplier No. MA553583
Only null left

Rabbit Recombinant Monoclonal Antibody

WNT5A Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.

Specifications

Antigen WNT5A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2400
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene WNT5A
Gene Accession No. P41221
Gene Alias 8030457G12Rik; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; Wnt family member 5A; WNT5A; Wnt-5a; WNT-5A protein
Gene Symbols WNT5A
Host Species Rabbit
Immunogen A synthesized peptide derived from human Wnt5a (330-360AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7474
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less