missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Wizard™ Magnetic DNA Purification System for Food
Description
Includes
MagneSil Paramagnetic Particles; Lysis Buffer A, Food; Lysis Buffer B, Food; Precipitation Solution, Food; RNase A Solution
Requires
MagneSphere Technology Magnetic Separation Stand (Z5332 or Z5342)
- Rapid Isolation of DNA from Many Food Types
- Requires minimal centrifugation and eliminates organic extractions
- Purified DNA suitable for PCR-based GMO testing
U.S. Patents 6,027,945 and 6,368,800, Australian Patent 732756 and other patents and patents pending.
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Application) | Purifies DNA from a variety of food samples including corn seeds, cornmeal, soybeans, Soy flour and Soy milk |
| Includes | 100mL Lysis Buffer A, 100mL Lysis Buffer B, 1mL RNase A, 19mL Magnesil™ paramagnetic particles, 150mL precipitation solution |
| No. of Reactions | 200 Preps |
| Product Type | System |